
Ammonium hydrogen carbonate, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- CHMP421403709.0250421403709.0250Glass bottle for solids250 gLog in for availability1 * 250 g32,90 zł
- CHMP421403709.0500421403709.0500Glass bottle for solids500 gLog in for availability1 * 500 g35,70 zł
- CHMP421403709.1000421403709.1000Glass bottle for solids1 kgLog in for availability1 * 1 kg49,60 zł