Drukuj
Regulator energii do łaźni
Dostawca: Nickel-Electro
452-0013EA 1490 PLN
452-0013
Regulator energii do łaźni
Zasilacze
This compact energy regulator is ideal for use with electric round and rectangular boiling baths excluding the 110-ER models. Suitable for 230 V, 50 to 60 Hz operation up to 10 amps.
  • Allows the boiling to be controlled from vigorous to gentle simmer
Informacje do zamówienia: Supplied with mains inlet and outlet leads.