Drukuj
Fiolki do przechowywania, opakowania kombinowane, Chromacol™
Dostawca: Thermo Fisher Scientific
Chromacol™
CRMA16-SVA-CPEA 0 PLN
CRMA16-SVA-CP
Fiolki do przechowywania, opakowania kombinowane, Chromacol™
Vialki
These storage vials are made of class 1 hydrolytic extraction neutral borosilicate glass to provide sample stability and low extraction.
Dodatkowe informacje: When using in automated storage systems the vials are supplied with pre-applied barcoded labels for specification. Both injection caps with piercable seals or solid PTFE lined storage caps are supplied separately, or in part of convenience packs.