Drukuj
Wyszukano: Odczynniki do oczyszczania białek
42  wyniki/ów wyszukiwania
SearchResultCount:"42"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: Avantor
Opis: Płytki J.T.Baker® BAKERBOND® do ekstrakcji wspomaganej cieczą (SLE) są idealną i szybszą alternatywą dla tradycyjnych metod ekstrakcji ciecz-ciecz i u...
Producent: Avantor
Opis: Płytki J.T.Baker® Bakerbond® do izolowania białek umożliwiają szybkie usuwanie białek z próbek biologicznych.
Numer katalogowy: (786-391)
Producent: G-Biosciences
Opis: Immobilised D-galactose resin
j.m.: 1 * 5 mL
Producent: G-Biosciences
Opis: Protein proteomic control, animal cells, 2×2 mg
Producent: G-Biosciences
Opis: Concanavalin A (Con A) agarose consists of Con A coupled to 6% agarose by the cyanogen bromide method. Con A is a tetrameric metalloprotein lectin iso...
Producent: G-Biosciences
Opis: Jacalin, immobilised, Typ: Liquid
Numer katalogowy: (786-243)
Producent: G-Biosciences
Opis: Following digestion of proteins with proteases, the addition of Pep-Extract™ rapidly elutes digested peptides in under 20 minutes. The eluted peptides...
j.m.: 1 * 100 Tests
Numer katalogowy: (PIER29997)
Producent: Thermo Fisher Scientific
Opis: Pierce™ Biotinylated Recombinant Protein L can be used to probe and detect mouse and human antibodies containing kappa light chains in Western blottin...
j.m.: 1 * 0,5 mg
MSDS
Numer katalogowy: (PIER32391)
Producent: Thermo Fisher Scientific
Opis: Pierce™ Recombinant Alkaline Phosphatase Conjugated Protein A/G is useful to probe and detect antibodies, especially IgG isotypes, of many different s...
j.m.: 1 * 0,5 mg
Numer katalogowy: (PIER77712)
Producent: Thermo Fisher Scientific
Opis: Immobilised TCEP disulphide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample fr...
j.m.: 1 * 5 mL
Numer katalogowy: (PIER77720)
Producent: Thermo Fisher Scientific
Opis: Bond-Breaker™ TCEP Solution (neutral pH) is a stable, 0,5 M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addit...
j.m.: 1 * 5 mL
MSDS
Producent: Thermo Fisher Scientific
Opis: Pierce™ MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) and TMS(PEG)12 is a ...
Producent: Thermo Fisher Scientific
Opis: Pierce™ High Sensitivity Streptavidin-HRP is a peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-...
Numer katalogowy: (PIER26099)
Producent: Thermo Fisher Scientific
Opis: Pierce™ SAT(PEG)₄ (PEGylated N-succinimidyl S-acetylthioacetate) is a sulphhydryl-addition reagent for covalent modification of primary amines.
j.m.: 1 * 100 mg
Numer katalogowy: (PIER44881)
Producent: Thermo Fisher Scientific
Opis: Pierce™ Immobilised Ficin consists of a sulfhydryl-specific protease (ficin) that has been immobilised onto beaded agarose resin to enable controlled ...
j.m.: 1 * 5 mL
Producent: Thermo Fisher Scientific
Opis: Purified (unconjugated) Pierce™ Recombinant Protein L is useful as the basis for preparing various kinds of probes or affinity media for detection or ...
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 42
no targeter for Bottom