Drukuj
Kolagen V
Dostawca: Corning
734-0106EA 2390 PLN
BDAA354246
Kolagen V
Collagen V is found in whole placenta, amnion, chorion, and cornea. It can be used as a thin coating on tissue culture surfaces to study collagen V effects on cell behavior. Collagen V has been shown to inhibit endothelial cell proliferation selectively.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma
Source material tested for hepatitis B antigen and HIV-1 antibody