Nickel sulphate heptahydrate, pure
Supplier: CHEMPUR
Certificates
About this item
Synonyms: Nickel (II) sulphate heptahydrate
CAS Number:10101-98-1
:
Formula:NiO₄S·7H₂O
MDL Number:MFCD00149814
Density:∼2 g/cm³ (20 °C)
Molecular Weight:280.86 g/mol
Melting Pt:99
EINECS:232-104-9
UN:3077
ADR:9 III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- CHMP426751508.0250426751508.0250Glass bottle for solids250 gLog in for availability1 * 250 g65 zł
- CHMP426751508.0500426751508.0500Glass bottle for solids500 gLog in for availability1 * 500 g95 zł
- CHMP426751508.1000426751508.1000Glass bottle for solids1 kgLog in for availability1 * 1 kg159 zł