Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00134045
Specifications
Related Information
Product Information
- Catalog #Supplier #ElementSymbolConcentrationMatrixPack typePkAvailabilityPrice
- 88066.AE88066.AEGalliumGa1000 mg/L5% HNO₃/tr HFGlass bottle100 mLLog in for availability1 * 100 mL420 złSupplier Article Number: 88066.AELocal Article Number: ALFA88066.AE
- 88066.AP88066.APGalliumGa1000 mg/L5% HNO₃/tr HFGlass bottle500 mLLog in for availability1 * 500 mL916 złSupplier Article Number: 88066.APLocal Article Number: ALFA88066.AP