Drukuj
Środki dezynfekcyjne, koncentrat, Mucocit®-T
Dostawca: Brand
BRND44822_BEA 964 PLN
BRND44822_B
Środki dezynfekcyjne, koncentrat, Mucocit®-T
Cleaning disinfectant fluid as a concentrate, particularly suited for sensitive instruments
  • Particularly suited for sensitive instruments
  • Free of phosphates, aldehydes, phenols and chlorine derivates
  • The solution has a pleasant, fresh fragrance
Disinfection of instruments: Bacteria (incl. Tuberculosis and mycobacteria) and fungi with high organic burden according to DGHM/VAH directives: 1% (10 g/L)/1 hour, 2% (20 g/L)/30 minutes, 3% (30 g/L/15 minutes. Limited virucidal efficacy* (incl. HIV, HBV, HCV): 1% (10 g/L)/5 minutes.Inactivation of human rotaviruses: 2% (20 g/L)/5 minutes
Certyfikaty: * As recommended by the RKI, Bundesgesundheitsblatt 01/2004.