Drukuj
System do drukowani i etykietowania kasetek osadczych, BSP™31
Dostawca: Brady Corporation
720-1968EA 17940 PLN
BRDY804855 BRDY804856 BRDY804859 BRDY804858
System do drukowani i etykietowania kasetek osadczych, BSP™31
A labelling system for histology and anatomic pathology specimen labelling. Two versions of the BSP™ Kit are available to meet both start-up requirements (Full) or expansion of existing operation (Light).
  • Clear, legible label information reduces identification errors
  • 2-D barcodes can be applied with small text for more information
  • Cassette and slide labels can be applied pre-process
  • Longevity of B-482 label is up to 20 years
The BSP™31 permanently attaches labels to tissue cassettes, resulting in more accurate specimen tracking and sample management. After a specimen label has been applied to a cassette, the cassette is placed in the labelling system. Four small heat probes pierce through the corners of the label and cassette. The heat melts the cassette's plastic into rivets, permanently holding the label in place. The specimen label remains identifiable through histology and anatomic pathology tissue processing protocols and up to 20 years in storage. For use with Brady® tissue cassette label material and thermal transfer printer ribbon.
Dodatkowe informacje: BSP™ Kit Full includes the compact bench thermal printer BBP™11, label attachment system BSP™31, a CR500 barcode reader, B-482 tissue cassette labels, B-481 StainerBondz™ slide labels together with R-6400 thermal transfer ribbon.
BSP™ Kit Light includes the compact bench thermal printer BBP™11 and the label attachment system BSP™31.