Cadmium(II) sulphate 8/3-hydrate, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7790-84-3
:
Formula:3CdSO₄·8H₂O
MDL Number:MFCD02253000
:256.51 g/mol (anhy)
Merck Index:13,01627
Melting Pt:41 °C
Density:3.79 g/cm³ (20 °C)
EINECS:233-331-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- CHMP114976705.0100114976705.0100Glass bottle for solids100 gLog in for availability1 * 100 g53,50 złSupplier Article Number: 114976705.0100Local Article Number: CHMP114976705.0100
- CHMP114976705.0250114976705.0250Plastic bottle for solids250 gLog in for availability1 * 250 g95,80 złSupplier Article Number: 114976705.0250Local Article Number: CHMP114976705.0250
- CHMP114976705.0500114976705.0500Glass bottle for solids500 gLog in for availability1 * 500 g135 złSupplier Article Number: 114976705.0500Local Article Number: CHMP114976705.0500
- CHMP114976705.1000114976705.1000Glass bottle for solids1 kgLog in for availability1 * 1 kg247 złSupplier Article Number: 114976705.1000Local Article Number: CHMP114976705.1000