Canada balsam for microscopy
Supplier: CHEMPUR
Certificates
About this item
CAS Number:8007-47-4
:
MDL Number:MFCD00132800
Merck Index:13,00948
Density:0.915 g/cm³ (20 °C)
Flash Pt:62 °C
Storage Temperature:Ambient
EINECS:232-362-2
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice