
Ethanol ≥96%, pure
POCH396420420500ML
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Assay:min. 96 %
- Density (20°C):max. 0.808 g/ml
- Refractive index (20°C):max. 1.364
- Acids (as CH₃COOH):max. 0.003 %
- Aldehydes (as CH₃CHO):max. 0.001 %
- Methanol:max. 0.1 %
- Higher alcohols (as amyl alcohol):max. 0.001 %
- Discolouration time of solution of KMnO₄:min. 5 min
- Miscibility with water:passes test
- Non-volatile residue:max. 0.001 %
- Furfurol presence test:passes test
- Ketones presence test:passes test
- Organic impurities:max. 0.5 %
- Heavy metals (as Pb):max. 0.0001 %
:
:46.07 g/mol
:–114 °C
:Ambient
:200-578-6
:1170
:3,II
:GHS02
:64-17-5
:C₂H₅OH
:MFCD00003568