Glycerine ≥85%, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:13,04497
Storage Temperature:Ambient
EINECS:200-289-5
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 424433204.0500Local Article Number: CHMP424433204.0500
- Supplier Article Number: 424433204.1000Local Article Number: CHMP424433204.1000