Sodium chloride 0.5 mol/l (0.5 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
:7647-14-5
:
:NaCl
:MFCD00003477
:58.44 g/mol
:Ambient
:231-598-3
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- CHMP817935104.0100817935104.0100Glass bottle100 mL1 * 100 mL47,40 złSupplier Article Number: 817935104.0100Local Article Number: CHMP817935104.0100
- CHMP817935104.0250817935104.0250Glass bottle250 mL1 * 250 mL55,70 złSupplier Article Number: 817935104.0250Local Article Number: CHMP817935104.0250
- CHMP817935104.0500817935104.0500Glass bottle500 mL1 * 500 mL64,10 złSupplier Article Number: 817935104.0500Local Article Number: CHMP817935104.0500
- CHMP817935104.1000817935104.1000Glass bottle1 L1 * 1 L106 złSupplier Article Number: 817935104.1000Local Article Number: CHMP817935104.1000