Drukuj
Wyszukano: Zestawy do testów środowiskowych
637  wyniki/ów wyszukiwania
SearchResultCount:"637"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Chemicals
Opis: A ready to use kit that fits into your lab coat pocket.
MSDS Certyfikaty Wyprzedaż
Numer katalogowy: (85708.601)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of sulphate.
j.m.: 1 * 100 SZT
Producent: VWR Chemicals
Opis: VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
Producent: VWR Chemicals
Opis: VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
Producent: VWR Chemicals
Opis: VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
Producent: VWR Chemicals
Opis: VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
Producent: VWR Chemicals
Opis: VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
MSDS Certyfikaty Wyprzedaż
Producent: VWR Chemicals
Opis: VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
Numer katalogowy: (85707.601)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of formaldehyde.
j.m.: 1 * 100 SZT
MSDS
Numer katalogowy: (85694.610)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of water hardness.
j.m.: 1 * 1.000 SZT
Numer katalogowy: (85600.620)
Producent: VWR Chemicals
Opis: VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
j.m.: 1 * 100 Tests
MSDS Certyfikaty
Numer katalogowy: (85620.620)
Producent: VWR Chemicals
Opis: VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary....
j.m.: 1 * 100 Tests
MSDS Certyfikaty
Numer katalogowy: (85710.601)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of arsenic.
j.m.: 1 * 100 SZT
MSDS
Numer katalogowy: (85697.601)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of nickel.
j.m.: 1 * 100 SZT
MSDS
Producent: VWR Chemicals
Opis: A ready to use kit that fits into your lab coat pocket.
Numer katalogowy: (85712.601)
Producent: VWR Chemicals
Opis: Semi-quantitative test strips for the determination of chlorine.
j.m.: 1 * 100 SZT
MSDS
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 637
no targeter for Bottom