Order Entry
Precision alcohol meters
Precision alcohol meters
Supplier:  Alla France
Certificates
undefined
Precision alcohol meters
Supplier:  Alla France
Product Information
  • Catalog #
    Supplier #
    Range
    Division
    Accuracy
    Length
    Pk
    Availability
    Price
  • Precision alcoholometer 40 - 100%, 0,5% vol, Tp. 20 °C
    Precision alcoholometer 40 - 100%, 0,5% vol, Tp. 20 °C
    ALLA0530FC040/20-Q
    0530FC040/20-Q
    40 - 100% EtOH
    0,5 %
    ± 0,5
    290 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 306 zł
    306 zł
    1 * 1 SZT
       306 zł
    Supplier Article Number: 0530FC040/20-Q
    Local Article Number: ALLA0530FC040/20-Q
    Specifications:
    Specifications
    Environmentally Preferable
  • Precision alcoholometer 0 - 50%, 0,5% vol., Tp. 20 °C
    Precision alcoholometer 0 - 50%, 0,5% vol., Tp. 20 °C
    ALLA0530FC050/20-Q
    0530FC050/20-Q
    50 - 100% EtOH
    0,5 %
    ± 0,5
    290 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 306 zł
    306 zł
    1 * 1 SZT
       306 zł
    Supplier Article Number: 0530FC050/20-Q
    Local Article Number: ALLA0530FC050/20-Q
    Specifications:
    Specifications
    Environmentally Preferable