Drukuj
Wycieraczki konserwacyjne, typ medium duty, PIG®
Dostawca: New Pig
PIG®
NEWPWIP216EA 0 PLN
NEWPWIP229 NEWPWIP216
Wycieraczki konserwacyjne, typ medium duty, PIG®
Chusteczki Chusteczki i ręczniki jednorazowe Chusteczki do ogólnych zastosowań
These nylon-scrim reinforced cellulose wipes are ideal for use in plants and workshops with brake fluid, antifreeze, grease, oils and more.
  • Indispensable in machine workshops, fleet garages and plants; unaffected by brake fluid, antifreeze, grease, oils and other liquids
  • Ideal for low-cost daily maintenance (not recommended for long-term contact with solvents)
  • Embossed surface grabs grime for tough cleaning with backup from 2-ply tissue and hidden nylon scrim
  • Low-lint material won't leave residue or create a mess
  • Available as an economical roll, 1/4-folded or in an easy-to-carry box that can be located conveniently at any workstation
  • Convenient pop-up box lets users take just one wipes at a time whilst keeping remaining stock clean and dry