Order Entry
Pig Pepsin (from Gastric mucosa)
Pig Pepsin (from Gastric mucosa)
Supplier:  Merck
Certificates
Pig Pepsin (from Gastric mucosa)
Supplier:  Merck
  • CAS No.:
    9001-75-6
  • Source:
    Gastric mucosa
  • Species:
    Pig
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pepsin
  • Enzyme activity:
    100 U/g
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • SIAL77151-1G
    77151-1G
    Glass bottle for solids
    1 g
    Log in for availability
    1 * 1 g
    1 * 1 g - 507 zł
    507 zł
    1 * 1 g
       507 zł
     : 77151-1G
     : SIAL77151-1G
  • SIAL77151-25G
    77151-25G
    Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 7 160 zł
    7 160 zł
    1 * 25 g
       7 160 zł
     : 77151-25G
     : SIAL77151-25G