Drukuj
Peroksydaza chrzanowa
Dostawca: MoBiTec
VWR®
MOBI0417100000UAMEA 0 PLN
MOBI0417100000UAM
Peroksydaza chrzanowa
Enzymy
HRP catalyses the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates.
Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.