Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
Enzymes
Horseradish peroxidase, Typ I
Horseradish peroxidase, Typ I
Horseradish peroxidase, Typ I
:
Merck
Horseradish peroxidase, Typ I
:
Merck
CAS No.:
9003-99-0
Species:
Horseradish
Storage conditions:
Freezer
MDL No.:
MFCD00071339
Enzyme name:
Peroxidase, Typ I
Pk
SIALP8125-5KU
P8125-5KU
5 KU
1 * 5 KU
1 * 5 KU - 289 zł
289 zł
-
+
1 * 5 KU
289 zł
-
+
SIALP8125-25KU
P8125-25KU
25 KU
1 * 25 KU
1 * 25 KU - 612 zł
612 zł
-
+
1 * 25 KU
612 zł
-
+
SIALP8125-100KU
P8125-100KU
100 KU
1 * 100 KU
1 * 100 KU - 1 920 zł
1 920 zł
-
+
1 * 100 KU
1 920 zł
-
+
SIALP8125-200KU
P8125-200KU
200 KU
1 * 200 KU
1 * 200 KU - 3 390 zł
3 390 zł
-
+
1 * 200 KU
3 390 zł
-
+