Measuring cylinders, conical
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
PP, translucent.
- Good chemical resistance
- Autoclavable to +121 °C
- Liquid volume is easy to read
Ideal for a wide range of applications. With moulded graduations. Broad base for stability.
Product Information
- Catalog #Supplier #CapacityDivisionHeightPkAvailabilityPrice
- HECH4225505442255054500 ml10 ml180 mm11 * 1 SZT69,50 zł: 42255054: HECH42255054
Specifications
Environmentally Preferable - : 42255048: HECH42255048
Specifications
Environmentally Preferable