Drukuj
Wialki z zamknięciem na zatrzask
Dostawca: Capitol Europe
216-0877EA 1730 PLN
216-0877 216-0876
Wialki z zamknięciem na zatrzask
Vialki
Aseptic, single-use sample vials made from translucent PP specifically designed to fit in the racks and transport carousels in use within Austria, Germany and Switzerland. Suitable for food contact.
  • Manufactured in a cleanroom
  • Proprietary "in mould closing" process
  • Can be supplied with optional RFID reusable tag
  • Can be supplied with automation concepts to insert and remove tags
MLP vials: Specific for quality and DHIA testing of milk. Ideally shaped for quality control and herd improvement purposes. Filling lines at 12,5, 25 and 37,5 ml. Can be supplied filled with bronopol tablet or bronopol gel. Can be supplied with barcode label on the top of the lid.
QK vials: Specific for quality testing of milk on automatic samplers. Ideally shaped for payment and quality control on automatic truck needle samplers
Certyfikaty: Complies with CE norm No.1935/2004
Sposób pakowania: A label on each box includes batch number and production date for traceability.
Uwaga: *Produkt(y) na tej stronie nie są objęte zakresem dyrektywy UE 2019/904