Order Entry
Centrifuge tubes, conical bottom
Centrifuge tubes, conical bottom
Supplier:  witeg Labortechnik
Certificates
undefined
Centrifuge tubes, conical bottom
Supplier:  witeg Labortechnik
Product Information
  • Catalog #
    Supplier #
    Capacity
    Ø×L
    Closure type
    Type
    Pk
    Availability
    Price
  • WITG4.947.002
    4.947.002
    35 ml
    28×100 mm
    Screw cap
    ungraduated
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 132 zł
    132 zł
    1 * 1 SZT
       132 zł
    Supplier Article Number: 4.947.002
    Local Article Number: WITG4.947.002
    Specifications:
    Specifications
    Environmentally Preferable
  • WITG4.947.001
    4.947.001
    10 ml
    20×97 mm
    Screw cap
    graduated
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 108 zł
    108 zł
    1 * 1 SZT
       108 zł
    Supplier Article Number: 4.947.001
    Local Article Number: WITG4.947.001
    Specifications:
    Specifications
    Environmentally Preferable