Order Entry
UltraSpeed cleaning system | Mops
UltraSpeed cleaning system
Supplier:  FHP VILEDA SCS
Certificates
undefined
UltraSpeed cleaning system
Supplier:  FHP VILEDA SCS
Handle
Product Information
  • Catalog #
    Supplier #
    Colour
    Dimensions
    Length
    Capacity
    Pk
    Availability
    Price
  • Telescopic handle
    Telescopic handle
    129-0807
    111389
    Grey/blue
    500 - 900 mm
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 184 zł
    184 zł
    1 * 1 SZT
       184 zł
    Accessories
    Specifications
    Environmentally Preferable
Mops and mops frame
  •  
    Colour
    Dimensions
    Length
    Capacity
    Pk
  • CombiSpeed 2 in1 mop frame: can be used with all commercial 40 cm and 50 cm pocket- and stripe mops
    CombiSpeed 2 in1 mop frame: can be used with all commercial 40 cm and 50 cm pocket- and stripe mops
    129-0793
    143572
    Grey/black
    40 cm
    1
    1 * 1 SZT
    1 * 1 SZT - 251 zł
    251 zł
    1 * 1 SZT
       251 zł
    Accessories
    Specifications
    Environmentally Preferable