Drukuj
Termocyklery, Techne® Prime
Dostawca: Cole Parmer
Techne®
732-1686EA 24360 PLN
TECN5PRIMEG/384 TECN5PRIMEX/USB TECN5PRIME/C TECN5PRIME/05 TECN5PRIME/384
Termocyklery, Techne® Prime
The Prime range of full-sized thermal cyclers delivers both high performance and high throughput to provide maximum flexibility when processing a large number of samples in parallel.
  • Colour touch screen for fast program set-up
  • Fast ramp rate of up to 3,4 °C/s
  • USB port enables the transfer of programs between instruments and temperature logs to your PC
  • Gradient cycling is available directly using the PrimeG instrument or by upgrading the Prime unit
The PrimeG is a gradient-enabled thermal cycler with all the features of the Prime unit. The wide linear gradient with a range of 30 °C allows annealing temperatures to be optimised in one experiment. The gradient calculator function displays the temperature for each of the eight columns, ensuring easy replication of thermal conditions.
Prime Combi-Block cyclers provide exceptional flexibility with the ability to accommodate 33×0,2 ml and 33×0,5 ml tubes, in 6 rows and 11 columns. Prime Combi-Block cyclers are available with or without a temperature gradient, which is upgradable later.
Note: Exchange of thermal cycler blocks must be carried out by a trained service engineer. Please contact your local Avantor sales office for details about replacement blocks available.
* Recorded at 55 °C