Order Entry
Centrifugal filters, Amicon® Ultra-0,5 ml
Centrifugal filters, Amicon® Ultra-0,5 ml
Supplier:  MILLIPORE
New Product
Certificates
undefined
Centrifugal filters, Amicon® Ultra-0,5 ml
Supplier:  MILLIPORE
New Product
Specifications
  • Catalog No:
  • MILYUFC500396
  • MILYUFC510024
  • MILYUFC503008
  • MILYUFC505024
  • MILYUFC500308
  • MILYUFC510008
  • MILYUFC5100BK
  • MILYUFC5003BK
  • MILYUFC505096
  • MILYUFC503096
  • MILYUFC5030BK
  • MILYUFC501096
  • MILYUFC510096
  • MILYUFC500324
  • MILYUFC501008
  • MILYUFC505008
  • MILYUFC5050BK
  • MILYUFC501024
  • MILYUFC503024
  • MILYUFC5010BK
  • Filtration area:
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • 1 cm²
  • Max. RCF:
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • 15000×g
  • Dead volume:
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • <1 µl
  • Ø×H:
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • 10,8×49,9 mm
  • Sample volume:
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
  • max. 500 µl
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    MWCO
    Membrane
    Colour code
    Filtration area
    Availability
    Price
  • MILYUFC500308
    UFC500308
    3 kD
    Ultracel-3, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 8 SZT
    1 * 8 SZT - 240 zł
    240 zł
    1 * 8 SZT
       240 zł
    Supplier Article Number: UFC500308
    Local Article Number: MILYUFC500308
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC500324
    UFC500324
    3 kD
    Ultracel-3, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 24 SZT
    1 * 24 SZT - 558 zł
    558 zł
    1 * 24 SZT
       558 zł
    Supplier Article Number: UFC500324
    Local Article Number: MILYUFC500324
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC500396
    UFC500396
    3 kD
    Ultracel-3, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 96 SZT
    1 * 96 SZT - 1 920 zł
    1 920 zł
    1 * 96 SZT
       1 920 zł
    Supplier Article Number: UFC500396
    Local Article Number: MILYUFC500396
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC5003BK
    UFC5003BK
    3 kD
    Ultracel-3, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 500 SZT
    1 * 500 SZT - 8 130 zł
    8 130 zł
    1 * 500 SZT
       8 130 zł
    Supplier Article Number: UFC5003BK
    Local Article Number: MILYUFC5003BK
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC501008
    UFC501008
    Ultracel-10, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 8 SZT
    1 * 8 SZT - 253 zł
    253 zł
    1 * 8 SZT
       253 zł
    Supplier Article Number: UFC501008
    Local Article Number: MILYUFC501008
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC501024
    UFC501024
    Ultracel-10, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 24 SZT
    1 * 24 SZT - 558 zł
    558 zł
    1 * 24 SZT
       558 zł
    Supplier Article Number: UFC501024
    Local Article Number: MILYUFC501024
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC501096
    UFC501096
    Ultracel-10, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 96 SZT
    1 * 96 SZT - 1 900 zł
    1 900 zł
    1 * 96 SZT
       1 900 zł
    Supplier Article Number: UFC501096
    Local Article Number: MILYUFC501096
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC5010BK
    UFC5010BK
    Ultracel-10, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 500 SZT
    1 * 500 SZT - 7 760 zł
    7 760 zł
    1 * 500 SZT
       7 760 zł
    Supplier Article Number: UFC5010BK
    Local Article Number: MILYUFC5010BK
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC503008
    UFC503008
    30 kD
    Ultracel-30, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 8 SZT
    1 * 8 SZT - 238 zł
    238 zł
    1 * 8 SZT
       238 zł
    Supplier Article Number: UFC503008
    Local Article Number: MILYUFC503008
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC503024
    UFC503024
    30 kD
    Ultracel-30, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 24 SZT
    1 * 24 SZT - 550 zł
    550 zł
    1 * 24 SZT
       550 zł
    Supplier Article Number: UFC503024
    Local Article Number: MILYUFC503024
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC503096
    UFC503096
    30 kD
    Ultracel-30, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 96 SZT
    1 * 96 SZT - 1 860 zł
    1 860 zł
    1 * 96 SZT
       1 860 zł
    Supplier Article Number: UFC503096
    Local Article Number: MILYUFC503096
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC5030BK
    UFC5030BK
    30 kD
    Ultracel-30, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 500 SZT
    1 * 500 SZT - 7 690 zł
    7 690 zł
    1 * 500 SZT
       7 690 zł
    Supplier Article Number: UFC5030BK
    Local Article Number: MILYUFC5030BK
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC505008
    UFC505008
    50 kD
    Ultracel-50, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 8 SZT
    1 * 8 SZT - 236 zł
    236 zł
    1 * 8 SZT
       236 zł
    Supplier Article Number: UFC505008
    Local Article Number: MILYUFC505008
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC505024
    UFC505024
    50 kD
    Ultracel-50, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 24 SZT
    1 * 24 SZT - 550 zł
    550 zł
    1 * 24 SZT
       550 zł
    Supplier Article Number: UFC505024
    Local Article Number: MILYUFC505024
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC505096
    UFC505096
    50 kD
    Ultracel-50, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 96 SZT
    1 * 96 SZT - 1 870 zł
    1 870 zł
    1 * 96 SZT
       1 870 zł
    Supplier Article Number: UFC505096
    Local Article Number: MILYUFC505096
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC5050BK
    UFC5050BK
    50 kD
    Ultracel-50, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 500 SZT
    1 * 500 SZT - 7 950 zł
    7 950 zł
    1 * 500 SZT
       7 950 zł
    Supplier Article Number: UFC5050BK
    Local Article Number: MILYUFC5050BK
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC510008
    UFC510008
    100 kD
    Ultracel-100, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 8 SZT
    1 * 8 SZT - 236 zł
    236 zł
    1 * 8 SZT
       236 zł
    Supplier Article Number: UFC510008
    Local Article Number: MILYUFC510008
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC510024
    UFC510024
    100 kD
    Ultracel-100, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 24 SZT
    1 * 24 SZT - 550 zł
    550 zł
    1 * 24 SZT
       550 zł
    Supplier Article Number: UFC510024
    Local Article Number: MILYUFC510024
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC510096
    UFC510096
    100 kD
    Ultracel-100, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 96 SZT
    1 * 96 SZT - 1 860 zł
    1 860 zł
    1 * 96 SZT
       1 860 zł
    Supplier Article Number: UFC510096
    Local Article Number: MILYUFC510096
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product
  • MILYUFC5100BK
    UFC5100BK
    100 kD
    Ultracel-100, regenerated cellulose
    Clear
    1 cm²
    Log in for availability
    1 * 500 SZT
    1 * 500 SZT - 8 010 zł
    8 010 zł
    1 * 500 SZT
       8 010 zł
    Supplier Article Number: UFC5100BK
    Local Article Number: MILYUFC5100BK
    Specifications:
    New Product
    More
    Accessories
    Specifications
    Environmentally Preferable
    Filtration area
    1 cm²
    Max. RCF
    15000×g
    Dead volume
    <1 µl
    Ø×H
    10,8×49,9 mm
    Sample volume
    max. 500 µl
    New Product