About this item
Fast-Freeze® flasks for FreeZone® freeze dry systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.
- Compatible with all major brands of freeze drying equipment, including labconco and Virtis™
Fast-Freeze® flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples.
Flat bottom, high-strength borosilicate glass flask withstands high heat and deep vacuum. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide-mouth design provides full access to entire inner diameter of glassware for easy loading and cleaning. Silicone rubber snap-on tops provide reliable high-vacuum seals. Flasks should be filled no more than one-third of the volume so a maximum surface area is achieved and efficient lyophilization is assured.
Flask adapters attach to Fast-Freeze® flasks for connection to valve ports on drying chambers or manifolds for freeze dry systems. Choose borosilicate glass or stainless steel, straight or 45° angle adapters in 0,63 cm (1/4") and 1,9 cm (3/4") diameters.
Flask holders help to prevent sample melt back. They allow a small fast freeze flask bottom containing the sample to be placed inside a larger fast freeze flask so that the vacuum surrounding the sample insulates it from the room atmosphere. Stainless steel. Flasks not included.
Specifications
- Catalog No:
- 531-0948
- 535-0113
- 535-0122
- LBCP7543800
- 535-0112
- 535-0168
- 531-0176
- 535-0121
- 535-0042
- 535-0123
- 535-0192
- 535-0046
- 535-0170
- 535-0045
- 535-0169
- 535-0044
- 535-0172
- 535-0043
- 535-0171
- 535-0125
- 535-0124
- LBCP7205100
- 535-0127
- LBCP7205101
- 535-0126
- 535-0131
- 535-0129
- LBCP7541101
- 535-0128
- 535-0132
- 531-0950
- 535-0130
- 535-0154
- LBCP7542901
- 535-0153
- 535-0146
- 535-0142
- 535-0048
- LBCP7538400
- 535-0135
- 535-0148
- 535-0047
- 535-0147
- 535-0150
- 535-0050
- 535-0149
- 535-0152
- 535-0049
- 535-0151
- 535-0114
- 535-0116
- 535-0115
- 535-0118
- 535-0117
- 535-0120
- 535-0119
- LBCP7542601
- LBCP7542701
- LBCP7207400
- LBCP7542801
- 535-0143
- 535-0213
- 535-0141
- 535-0145
- 535-0144
- LBCP7121725
- LBCP7121724
- LBCP7569000
- 535-0182
- 535-0136
- 535-0181
- 535-0134
- 535-0176
- 535-0138
- 535-0137
- 535-0140
- 535-0139
- LBCP7426400
- LBCP7540801
- LBCP7540901
- 531-0941
- LBCP7121722
- LBCP7121721
- LBCP7121723
- LBCP7121711
- LBCP7121702
- LBCP7728000
- LBCP7121713
- LBCP7121712
- LBCP7121701
- LBCP7540601
- LBCP7509200
- LBCP7201700
- 535-0110
- LBCP7509401
- LBCP7509400
- 535-0214
- 535-0109
- 535-0108
- 535-0187
- 535-0167
- LBCP7645900
- LBCP7543400
- 535-0157
- 535-0186
- 535-0156
- LBCP7578100
- LBCP7578000
- LBCP7578500
- 535-0160
- 535-0159
- LBCP7317100
- 535-0162
- 535-0161
- 535-0164
- 535-0163
- 535-0166
- LBCP7394800
- 535-0165
- 535-0158
- 535-0184
- 535-0183
- 535-0111
- 535-0133
- 535-0178
- 531-0177
- 535-0174
- 535-0173
- 535-0177
- 535-0175
- 535-0180
- 535-0179
- 535-0191
- LBCP7207500
- 535-0185
- 535-0155
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Materials, Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Materials
- Sustainable Materials, Sustainable Packaging
Specifications
Fast-Freeze® flasks (complete set), and components
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 40 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 40 ml535-004275400001Log in for availability1 * 1 SZT669 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 80 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 80 ml531-017675402001Log in for availability1 * 1 SZT944 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 120 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 120 ml535-011275403001Log in for availability1 * 1 SZT1 290 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 150 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 150 ml535-004375404001Log in for availability1 * 1 SZT1 330 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 300 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 300 ml535-004475406001Log in for availability1 * 1 SZT1 560 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 600 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 600 ml535-004575408001Log in for availability1 * 1 SZT1 950 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 900 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 900 ml535-004675409001Log in for availability1 * 1 SZT2 090 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 1200 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 1200 ml535-011375410001Log in for availability1 * 1 SZT2 340 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 750 ml flask, H×D: 162×90,2 mmFast Freeze® flask bottom, glass, for 750 ml flask, H×D: 162×90,2 mm535-012175427001Log in for availability1 * 1 SZT1 250 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 600 ml flask, H×D: 135×90,2 mmFast Freeze® flask bottom, glass, for 600 ml flask, H×D: 135×90,2 mm535-012275428001Log in for availability1 * 1 SZT1 050 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 900 ml flask, H×D: 190×90,2 mmFast Freeze® flask bottom, glass, for 900 ml flask, H×D: 190×90,2 mm535-012375429001Log in for availability1 * 1 SZT1 320 złSpecifications:
Specifications
Environmentally Preferable - Adapters, stainless steel, for 29/32 round bottom flasks to valve portsAdapters, stainless steel, for 29/32 round bottom flasks to valve portsLBCP754380075438001Log in for availability1 * 1 SZT403 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask, 750 ml, amber.Neck diameter is 19 cmFast freeze flask, 750 ml, amber.Neck diameter is 19 cmLBCP754110175411011Log in for availability1 * 1 SZT2 070 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask bottom, 900 ml, amber.Neck diameter is 19 cmFast freeze flask bottom, 900 ml, amber.Neck diameter is 19 cmLBCP754290175429011Log in for availability1 * 1 SZT1 460 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 1200 ml flask, H×D: 240×90,2 mmFast Freeze® flask bottom, glass, for 1200 ml flask, H×D: 240×90,2 mm535-012475430001Log in for availability1 * 1 SZT1 530 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 2000 ml flask, H×D: 380×90,2 mmFast Freeze® flask bottom, glass, for 2000 ml flask, H×D: 380×90,2 mm535-012575432001Log in for availability1 * 1 SZT1 950 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask top, silicone rubber, for 120, 150 and 300 ml flasks,Fast Freeze® flask top, silicone rubber, for 120, 150 and 300 ml flasks,535-012775442001Log in for availability1 * 1 SZT380 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask top, silicone rubber, for 40 and 80 ml flasks,Fast Freeze® flask top, silicone rubber, for 40 and 80 ml flasks,535-012675440001Log in for availability1 * 1 SZT265 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask top, silicone rubber, for 600, 750, 900, 1200 and 2000 ml flasksFast Freeze® flask top, silicone rubber, for 600, 750, 900, 1200 and 2000 ml flasks535-012875444001Log in for availability1 * 1 SZT720 złSpecifications:
Specifications
Environmentally Preferable - Replacement filter paper of Ø 28 mm, poresize 17 micronsReplacement filter paper of Ø 28 mm, poresize 17 microns535-012975448101.000Log in for availability1 * 1.000 SZT811 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, straight, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve portsAdapters, borosilicate glass, straight, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve ports535-013075450001Log in for availability1 * 1 SZT94,90 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, straight, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve portsAdapters, borosilicate glass, straight, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve ports535-013175452001Log in for availability1 * 1 SZT121 złSpecifications:
Specifications
Environmentally Preferable - Adapters ¹/₂ - ³/₄", borosilicate glass, straight, for connecting the 12,7 and 19,05 mm Fast-Freeze® flask tops to 19,05 and 12,7 mm valve ports respectively.Adapters ¹/₂ - ³/₄", borosilicate glass, straight, for connecting the 12,7 and 19,05 mm Fast-Freeze® flask tops to 19,05 and 12,7 mm valve ports respectively.535-013275454001Log in for availability1 * 1 SZT170 złSpecifications:
Specifications
Environmentally Preferable - Adapters, stainless steel, 45° bend, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve portsAdapters, stainless steel, 45° bend, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve ports535-004875476001Log in for availability1 * 1 SZT309 złSpecifications:
Specifications
Environmentally Preferable - Adapters, stainless steel, straight, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve portsAdapters, stainless steel, straight, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve ports535-013575472001Log in for availability1 * 1 SZT237 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, 45° bend, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve portsAdapters, borosilicate glass, 45° bend, for connecting the 19,05 mm Fast-Freeze® flask top to 19,05 mm valve ports535-004775458001Log in for availability1 * 1 SZT168 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 750 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 750 ml535-011475411001Log in for availability1 * 1 SZT1 970 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® complete flask (incl. flask top, bottom and filter paper), 2000 mlFast Freeze® complete flask (incl. flask top, bottom and filter paper), 2000 ml535-011575412001Log in for availability1 * 1 SZT2 770 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 40 ml flask, H×D: 76×34 mmFast Freeze® flask bottom, glass, for 40 ml flask, H×D: 76×34 mm535-011675420001Log in for availability1 * 1 SZT510 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 80 ml flask, H×D: 115×34 mmFast Freeze® flask bottom, glass, for 80 ml flask, H×D: 115×34 mm535-011775422001Log in for availability1 * 1 SZT648 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 120 ml flask, H×D: 68×59,2 mmFast Freeze® flask bottom, glass, for 120 ml flask, H×D: 68×59,2 mm535-011875423001Log in for availability1 * 1 SZT811 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 150 ml flask, H×D: 85×59,2 mmFast Freeze® flask bottom, glass, for 150 ml flask, H×D: 85×59,2 mm535-011975424001Log in for availability1 * 1 SZT852 złSpecifications:
Specifications
Environmentally Preferable - Fast Freeze® flask bottom, glass, for 300 ml flask, H×D: 145×59,2 mmFast Freeze® flask bottom, glass, for 300 ml flask, H×D: 145×59,2 mm535-012075426001Log in for availability1 * 1 SZT959 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask bottom, 300 ml, amber.Neck diameter is 19 cmFast freeze flask bottom, 300 ml, amber.Neck diameter is 19 cmLBCP754260175426011Log in for availability1 * 1 SZT908 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask Bottom, 750 ml, amber.Neck diameter is 19 cmFast freeze flask Bottom, 750 ml, amber.Neck diameter is 19 cmLBCP754270175427011Log in for availability1 * 1 SZT1 340 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask bottom, 600 ml, amber.Neck diameter is 19 cmFast freeze flask bottom, 600 ml, amber.Neck diameter is 19 cmLBCP754280175428011Log in for availability1 * 1 SZT1 290 złSpecifications:
Specifications
Environmentally Preferable - Adapters, stainless steel, 45° bend, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve portsAdapters, stainless steel, 45° bend, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve ports535-013675474001Log in for availability1 * 1 SZT309 złSpecifications:
Specifications
Environmentally Preferable - Adapters, stainless steel, straight, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve portsAdapters, stainless steel, straight, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve ports535-013475470001Log in for availability1 * 1 SZT237 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask, 600 ml, amber.Neck diameter is 19 cmFast freeze flask, 600 ml, amber.Neck diameter is 19 cmLBCP754080175408011Log in for availability1 * 1 SZT1 970 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask, 900 ml, amber.Neck diameter is 19 cmFast freeze flask, 900 ml, amber.Neck diameter is 19 cmLBCP754090175409011Log in for availability1 * 1 SZT2 170 złSpecifications:
Specifications
Environmentally Preferable - Fast freeze flask, 300 ml, amber. Neck diameter is 19 cmFast freeze flask, 300 ml, amber. Neck diameter is 19 cmLBCP754060175406011Log in for availability1 * 1 SZT1 610 złSpecifications:
Specifications
Environmentally Preferable - Adapters ¹/₂ - ³/₄" angled, borosilicate glass, 45° bend, for connecting the 12,7 and 19,05 mm Fast-Freeze® flask tops to 19,05 and 12,7 mm valve ports respectively.Adapters ¹/₂ - ³/₄" angled, borosilicate glass, 45° bend, for connecting the 12,7 and 19,05 mm Fast-Freeze® flask tops to 19,05 and 12,7 mm valve ports respectively.535-013375460001Log in for availability1 * 1 SZT271 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, 45° bend, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve portsAdapters, borosilicate glass, 45° bend, for connecting the 12,7 mm Fast-Freeze® flask top to 12,7 mm valve ports531-017775456001Log in for availability1 * 1 SZT168 złSpecifications:
Specifications
Environmentally Preferable
Lyph-Lock® flasks (complete set), and components
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Adapters, borosilicate glass, 90° bend, for connecting the 24/40 STJ Lyph-Lock® flask Top to 19,05 mm valveAdapters, borosilicate glass, 90° bend, for connecting the 24/40 STJ Lyph-Lock® flask Top to 19,05 mm valve535-016875686001Log in for availability1 * 1 SZT411 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 20/40 STJ flasks (100 and 250 ml)Lyph-Lock® flask top, glass, for 20/40 STJ flasks (100 and 250 ml)535-015475562001Log in for availability1 * 1 SZT1 180 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 750 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 750 ml535-014275509001Log in for availability1 * 1 SZT3 140 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 25 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 25 ml535-014675540001Log in for availability1 * 1 SZT1 880 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 20/40 STJ flasks (25 and 50 ml)Lyph-Lock® flask top, glass, for 20/40 STJ flasks (25 and 50 ml)535-015375560001Log in for availability1 * 1 SZT791 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 500 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 500 ml535-014175508001Log in for availability1 * 1 SZT3 140 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 1000 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 1000 ml535-004975510001Log in for availability1 * 1 SZT3 380 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 19/38 STJ flasks (25 and 50 ml)Lyph-Lock® flask top, glass, for 19/38 STJ flasks (25 and 50 ml)535-014375520001Log in for availability1 * 1 SZT725 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 19/38 STJ flasks (500, 750 and 1000 ml)Lyph-Lock® flask top, glass, for 19/38 STJ flasks (500, 750 and 1000 ml)535-014575524001Log in for availability1 * 1 SZT1 150 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 50 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 50 ml535-014775542001Log in for availability1 * 1 SZT2 260 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 100 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 100 ml535-014875544001Log in for availability1 * 1 SZT2 550 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 19/38 STJ flasks (100 and 250 ml)Lyph-Lock® flask top, glass, for 19/38 STJ flasks (100 and 250 ml)535-014475522001Log in for availability1 * 1 SZT1 030 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 250 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 250 ml535-014975546001Log in for availability1 * 1 SZT2 650 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 500 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 500 ml535-015075548001Log in for availability1 * 1 SZT3 040 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 750 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 750 ml535-015175549001Log in for availability1 * 1 SZT3 140 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 1000 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 24/40 STJ, 1000 ml535-015275550001Log in for availability1 * 1 SZT3 380 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, 90° bend, for connecting the 19/38 STJ Lyph-Lock® flask top to 12,7 mm valveAdapters, borosilicate glass, 90° bend, for connecting the 19/38 STJ Lyph-Lock® flask top to 12,7 mm valve535-005075680001Log in for availability1 * 1 SZT380 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 25 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 25 ml535-013775500001Log in for availability1 * 1 SZT2 170 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 50 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 50 ml535-013875502001Log in for availability1 * 1 SZT2 210 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 100 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 100 ml535-013975504001Log in for availability1 * 1 SZT2 260 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 250 mlLyph-Lock® complete flask (incl. flask top, bottom and seal), 19/38 STJ, 250 ml535-014075506001Log in for availability1 * 1 SZT2 430 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, 90° bend, for connecting the 24/40 STJ Lyph-Lock® flask Top to 12,7 mm valveAdapters, borosilicate glass, 90° bend, for connecting the 24/40 STJ Lyph-Lock® flask Top to 12,7 mm valve535-016775684001Log in for availability1 * 1 SZT411 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 50 ml flask, H×D: 67×34 mmLyph-Lock® flask bottom, glass, for 50 ml flask, H×D: 67×34 mm535-015775572001Log in for availability1 * 1 SZT435 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 25 ml flask, H×D: 37×34 mmLyph-Lock® flask bottom, glass, for 25 ml flask, H×D: 37×34 mm535-015675570001Log in for availability1 * 1 SZT412 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 250 ml flask, H×D: 110×59,2 mmLyph-Lock® flask bottom, glass, for 250 ml flask, H×D: 110×59,2 mm535-015975576001Log in for availability1 * 1 SZT546 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 500 ml flask, H×D: 103×90,2 mmLyph-Lock® flask bottom, glass, for 500 ml flask, H×D: 103×90,2 mm535-016075578001Log in for availability1 * 1 SZT602 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 100 ml flask, H×D: 50×59,2 mmLyph-Lock® flask bottom, glass, for 100 ml flask, H×D: 50×59,2 mm535-015875574001Log in for availability1 * 1 SZT510 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 750 ml flask, H×D: 145×90,2 mmLyph-Lock® flask bottom, glass, for 750 ml flask, H×D: 145×90,2 mm535-016175579001Log in for availability1 * 1 SZT714 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask bottom, glass, for 1000 ml flask, H×D: 187×90,2 mmLyph-Lock® flask bottom, glass, for 1000 ml flask, H×D: 187×90,2 mm535-016275580001Log in for availability1 * 1 SZT760 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask seal, rubber, for 25 and 50 ml flasks of 19/38 and 24/40 STJLyph-Lock® flask seal, rubber, for 25 and 50 ml flasks of 19/38 and 24/40 STJ535-016375590001Log in for availability1 * 1 SZT633 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask seal, rubber, for 100 and 250 ml flasks of 19/38 and 24/40 STJLyph-Lock® flask seal, rubber, for 100 and 250 ml flasks of 19/38 and 24/40 STJ535-016475592001Log in for availability1 * 1 SZT602 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask seal, rubber, for 500, 750 and 1000 ml flasks of 19/38 and 24/40 STJLyph-Lock® flask seal, rubber, for 500, 750 and 1000 ml flasks of 19/38 and 24/40 STJ535-016575594001Log in for availability1 * 1 SZT694 złSpecifications:
Specifications
Environmentally Preferable - Adapters, borosilicate glass, 90° bend, for connecting the 19/38 STJ Lyph-Lock® flask Top to 19,05 mm valveAdapters, borosilicate glass, 90° bend, for connecting the 19/38 STJ Lyph-Lock® flask Top to 19,05 mm valve535-016675682001Log in for availability1 * 1 SZT380 złSpecifications:
Specifications
Environmentally Preferable - Lyph-Lock® flask top, glass, for 20/40 STJ flasks (500, 750 and 1000 ml)Lyph-Lock® flask top, glass, for 20/40 STJ flasks (500, 750 and 1000 ml)535-015575564001Log in for availability1 * 1 SZT1 290 złSpecifications:
Specifications
Environmentally Preferable
Serum bottles, vials and components
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Serum bottle, 3 ml, with 13 mm corkageSerum bottle, 3 ml, with 13 mm corkage535-01827575210100Log in for availability1 * 100 SZT1 150 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 2 ml, with 13 mm corkageSerum bottle, 2 ml, with 13 mm corkage535-01817575010100Log in for availability1 * 100 SZT990 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 20 ml, with 20 mm corkageSerum bottle, 20 ml, with 20 mm corkage535-01767573410100Log in for availability1 * 100 SZT1 880 złSpecifications:
Specifications
Environmentally Preferable - Seal, aluminium for 5, 10, 20, 30, 50, 100 and 125 ml serum bottles with 20 mm stoppersSeal, aluminium for 5, 10, 20, 30, 50, 100 and 125 ml serum bottles with 20 mm stoppers535-01867577110100Log in for availability1 * 100 SZT111 złSpecifications:
Specifications
Environmentally Preferable - Split stopper 13 mm, rubber for 2 and 3 ml serum bottlesSplit stopper 13 mm, rubber for 2 and 3 ml serum bottles535-01837576010100Log in for availability1 * 100 SZT242 złSpecifications:
Specifications
Environmentally Preferable - Split stopper 20 mm, rubber, for 5, 10, 20, 30, 50, 100 and 125 ml serum bottlesSplit stopper 20 mm, rubber, for 5, 10, 20, 30, 50, 100 and 125 ml serum bottles535-01847576210100Log in for availability1 * 100 SZT262 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 50 ml, with 20 mm corkageSerum bottle, 50 ml, with 20 mm corkage535-01787573810100Log in for availability1 * 100 SZT3 380 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 5 ml, with 20 mm corkageSerum bottle, 5 ml, with 20 mm corkage535-01747573010100Log in for availability1 * 100 SZT1 300 złSpecifications:
Specifications
Environmentally Preferable - Seal, aluminium for 2 and 3 ml serum bottles with 13 mm stoppersSeal, aluminium for 2 and 3 ml serum bottles with 13 mm stoppers535-01857577010100Log in for availability1 * 100 SZT110 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 10 ml, with 20 mm corkageSerum bottle, 10 ml, with 20 mm corkage535-01757573210100Log in for availability1 * 100 SZT1 320 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 30 ml, with 20 mm corkageSerum bottle, 30 ml, with 20 mm corkage535-01777573610100Log in for availability1 * 100 SZT1 920 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 100 ml, with 20 mm corkageSerum bottle, 100 ml, with 20 mm corkage535-01797574010100Log in for availability1 * 100 SZT7 500 złSpecifications:
Specifications
Environmentally Preferable - Serum bottle, 125 ml, with 20 mm corkageSerum bottle, 125 ml, with 20 mm corkage535-01807574210100Log in for availability1 * 100 SZT8 270 złSpecifications:
Specifications
Environmentally Preferable
Ampules and ampule adapters
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Ampoule, pre-scored flat bottom, borosilicate glass, 2 mlAmpoule, pre-scored flat bottom, borosilicate glass, 2 ml535-01707570810100Log in for availability1 * 100 SZT2 140 złSpecifications:
Specifications
Environmentally Preferable - Ampoule, pre-scored flat bottom, borosilicate glass, 1 mlAmpoule, pre-scored flat bottom, borosilicate glass, 1 ml535-01697570610100Log in for availability1 * 100 SZT1 880 złSpecifications:
Specifications
Environmentally Preferable - Ampoule, pre-scored flat bottom, borosilicate glass, 10 mlAmpoule, pre-scored flat bottom, borosilicate glass, 10 ml535-01727571210100Log in for availability1 * 100 SZT3 380 złSpecifications:
Specifications
Environmentally Preferable - Ampoule, pre-scored flat bottom, borosilicate glass, 5 mlAmpoule, pre-scored flat bottom, borosilicate glass, 5 ml535-01717571010100Log in for availability1 * 100 SZT2 920 złSpecifications:
Specifications
Environmentally Preferable - Ampoule valve adapterSpecifications:
Specifications
Environmentally Preferable - Ampoule, straight, borosilicate glass, 1 mlAmpoule, straight, borosilicate glass, 1 ml535-01737572710100Log in for availability1 * 100 SZT4 060 złSpecifications:
Specifications
Environmentally Preferable - Ampoule valve adapter, three waySpecifications:
Specifications
Environmentally Preferable
Other accessories
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Mini Stoppering Chamber, attaches to a port on a Labconco drying chamber or manifold (sold separately), to stopper small vials (up to 30 ml each), includes 432 mm of clear PVC tubingMini Stoppering Chamber, attaches to a port on a Labconco drying chamber or manifold (sold separately), to stopper small vials (up to 30 ml each), includes 432 mm of clear PVC tubingLBCP756900075690001Log in for availability1 * 1 SZT9 800 złSpecifications:
Specifications
Environmentally Preferable - Replacement cartridge for soda acid trapSpecifications:
Specifications
Environmentally Preferable - Attachment port lid, 264 mm diameterSpecifications:
Specifications
Environmentally Preferable - Ampoule pod 1/4" internal Ø tubing. Attaches to a single valve port and accommodates up to 15 ampoules for simultaneous freeze drying. For ampoule neck sizes less than 10 mm. Includes 15 plugs, 2 clamps and 1 straight stainless steel adapter.Ampoule pod 1/4" internal Ø tubing. Attaches to a single valve port and accommodates up to 15 ampoules for simultaneous freeze drying. For ampoule neck sizes less than 10 mm. Includes 15 plugs, 2 clamps and 1 straight stainless steel adapter.LBCP720510072051001Log in for availability1 * 1 SZT4 470 złSpecifications:
Sustainable Packaging More
Specifications
Environmentally PreferableSustainable PackagingSustainable Packaging - Ampoule pod 3/8" internal Ø tubing. Attaches to a single valve port and accommodates up to 15 ampoules for simultaneous freeze drying. For ampoule neck sizes larger than 10 mm. Includes 15 plugs, 2 clamps and 1 straight stainless steel adapter.Ampoule pod 3/8" internal Ø tubing. Attaches to a single valve port and accommodates up to 15 ampoules for simultaneous freeze drying. For ampoule neck sizes larger than 10 mm. Includes 15 plugs, 2 clamps and 1 straight stainless steel adapter.LBCP720510172051011Log in for availability1 * 1 SZT4 470 złSpecifications:
Sustainable Packaging More
Specifications
Environmentally PreferableSustainable PackagingSustainable Packaging - Replacement cartridge for activated carbon trapSpecifications:
Specifications
Environmentally Preferable - Dry ice secondary vacuum trap, Ø×h 167×198 mm, weight 2,26 kgDry ice secondary vacuum trap, Ø×h 167×198 mm, weight 2,26 kgLBCP753840075384001Log in for availability1 * 1 SZT8 010 złSpecifications:
Specifications
Environmentally Preferable - FreeZone® Stainless Steel Lid, Ø 355,6 mm, FreeZone 4,5, 6, 8, 12, and 18 L for use with freeze dryersFreeZone® Stainless Steel Lid, Ø 355,6 mm, FreeZone 4,5, 6, 8, 12, and 18 L for use with freeze dryersLBCP720740072074001Log in for availability1 * 1 SZT1 800 złSpecifications:
Specifications
Environmentally Preferable - Glass lid, for 4,5 l systems, Ø 257 mmSpecifications:
Specifications
Environmentally Preferable - Mesh sleeve for 1200 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Stainless steel lid, for 1, 2,5, and 4,5 l systems, Ø 257 mmStainless steel lid, for 1, 2,5, and 4,5 l systems, Ø 257 mm531-094173868001Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Support shelves, W×D 187×304 mm, weight of 0,45 kgSupport shelves, W×D 187×304 mm, weight of 0,45 kgLBCP742640074264002Log in for availability1 * 2 SZTSpecifications:
Specifications
Environmentally Preferable - 4-Port manifold, 304 stainless steel with four neoprene valves and latching pin, accomodates both 1/2” and 3/4” adapters for connecting flasks4-Port manifold, 304 stainless steel with four neoprene valves and latching pin, accomodates both 1/2” and 3/4” adapters for connecting flasksLBCP720170072017001Log in for availability1 * 1 SZT6 180 złSpecifications:
Sustainable Materials Sustainable Packaging More
Specifications
Environmentally PreferableSustainable Materials, Sustainable PackagingSustainable Materials Sustainable Packaging - Tube holder, stainless steel, holds 15 tubes of 12 or 13 mm Ø tubesTube holder, stainless steel, holds 15 tubes of 12 or 13 mm Ø tubes535-010874426001Log in for availability1 * 1 SZT1 830 złSpecifications:
Sustainable Packaging More
Specifications
Environmentally PreferableSustainable PackagingSustainable Packaging - Tube holder, stainless steel, holds 30 tubes of 2 ml microcentrifuge tubesTube holder, stainless steel, holds 30 tubes of 2 ml microcentrifuge tubes535-010974427001Log in for availability1 * 1 SZT1 830 złSpecifications:
Sustainable Packaging More
Specifications
Environmentally PreferableSustainable PackagingSustainable Packaging - Heated product shelf with variable heat control, 151×229 mm for use with drying chambersHeated product shelf with variable heat control, 151×229 mm for use with drying chambersLBCP750920075092001Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Heated product shelf with variable heat control, 159×229 mm for use with drying chambersHeated product shelf with variable heat control, 159×229 mm for use with drying chambers535-011075092011Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Heated product shelf with variable heat control, 245×298 mm for use with drying chambersHeated product shelf with variable heat control, 245×298 mm for use with drying chambersLBCP750940075094001Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Heated product shelf with variable heat control, 245×298 mm for use with drying chambersHeated product shelf with variable heat control, 245×298 mm for use with drying chambersLBCP750940175094011Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Replacement valve kit, includes neoprene body and gaskets, moulded plastic knob and fittings, accommodates both 1/2" and 3/4" adaptersReplacement valve kit, includes neoprene body and gaskets, moulded plastic knob and fittings, accommodates both 1/2" and 3/4" adapters535-021475096001Log in for availability1 * 1 SZT633 złSpecifications:
Specifications
Environmentally Preferable - Drain plug for Freeze dry systems, FreeZone 1 l, 2,5 l, 4,5 l, 6 l, 12 l and 18 lDrain plug for Freeze dry systems, FreeZone 1 l, 2,5 l, 4,5 l, 6 l, 12 l and 18 lLBCP772800077280001Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Mesh sleeve for 40 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 80 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 120 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 150 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 300 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 600 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 750 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Mesh sleeve for 900 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product - Large flask holder, for 120, 150 and 300 ml inner flasks and 750 and 1200 ml outer flasksLarge flask holder, for 120, 150 and 300 ml inner flasks and 750 and 1200 ml outer flasksLBCP754340075434001Log in for availability1 * 1 SZT416 złSpecifications:
Specifications
Environmentally Preferable - Drain hose, for 1 and 4,5 l systems, weight 0,2 kgDrain hose, for 1 and 4,5 l systems, weight 0,2 kgLBCP764590076459001Log in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - Removable shelves, small, aluminium, W×D×H: 277×264×249 mm, weight: 2,5 kg for use with drying chambersRemovable shelves, small, aluminium, W×D×H: 277×264×249 mm, weight: 2,5 kg for use with drying chambersLBCP731710073171001Log in for availability1 * 1 SZT5 820 złSpecifications:
Sustainable Materials More
Specifications
Environmentally PreferableSustainable MaterialsSustainable Materials - Liquid nitrogen secondary trap, weight 10,8 kgLiquid nitrogen secondary trap, weight 10,8 kgLBCP739480073948001Log in for availability1 * 1 SZT34 840 złSpecifications:
Specifications
Environmentally Preferable - Seal crimper, for 13 mm corkageSpecifications:
Specifications
Environmentally Preferable - Seal crimper, for 20 mm corkageSpecifications:
Specifications
Environmentally Preferable - Organic/natural gas sealing torchSpecifications:
Specifications
Environmentally Preferable - 20-Port manifold, 304 stainless steel with twenty neoprene valves, accommodates both 1/2" and 3/4" adaptors for connection to flasks20-Port manifold, 304 stainless steel with twenty neoprene valves, accommodates both 1/2" and 3/4" adaptors for connection to flasks535-011175223001Log in for availability1 * 1 SZT20 100 złSpecifications:
Sustainable Materials Sustainable Packaging More
Specifications
Environmentally PreferableSustainable Materials, Sustainable PackagingSustainable Materials Sustainable Packaging - FreeZone® Stainless Steel Lid, Ø 256,54 mm, FreeZone 2,5 L for use with freeze dryersFreeZone® Stainless Steel Lid, Ø 256,54 mm, FreeZone 2,5 L for use with freeze dryersLBCP720750072075001Log in for availability1 * 1 SZT1 610 złSpecifications:
Specifications
Environmentally Preferable - Mesh sleeve for 2000 ml flaskSpecifications:New Product
Specifications
Environmentally PreferableNew Product