Drukuj
Stopcocks, high vacuum, safety pattern, single bore
Dostawca: SciLabware
BIBBSSV802EA 389 PLN
BIBBSSV802
Stopcocks, high vacuum, safety pattern, single bore
Stopcocks, made of PYREX® borosilicate glass, designed for the high vacuum performance.
  • Safety pattern and single-bore
  • Side-arm length of 100 mm
  • Holds the key firmly when connected to side-arm the bulb is evacuated
  • Prevents accidental removal of the key whilst the system is under vacuum