Human Recombinant GC-C (from HEK293 Cells)
New Product
- Koniugacja:Unconjugated
- Rodzaj białka / peptydu:Rekombinowany
- Źródło:HEK293 cells
- Gatunki:Human
- Warunki przechowywania:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Identyfikator genu:UniprotID# P25092
- Instrukcje dotyczące odtwarzania:Reconstitute with distilled sterile water
- Bez przewoźnika:N
- Synonimy białek:Guanylyl cyclase C|GUC2C|Heat-stable enterotoxin receptor|hSTAR|Intestinal guanylate cyclase|STA receptor
- Nazwa białka / peptydu:GC-C
- Czystość:>95% by SDS-PAGE and Coomassie blue staining
- Waga molekularna:The protein has a predicted molecular mass of 46,8 kDa after removal of the signal peptide The apparent molecular mass of GUCY2C-His is approximately 55 - 130 kDa due to glycosylation
- Sekwencja:GUCY2C (Ser24-Gln430) + 6×His Tag
- Stężenie:Reconstitution dependent
- Formulacja:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Temperatura transportu:Shipped on blue ice at 4 °C
- Przetestowane aplikacje:ELISA, SDS-PAGE
- Rodzaj opakowania
- : A318285-100: ANTIA318285-100New ProductNew Product
- : A318285-50: ANTIA318285-50New ProductNew Product