Order Entry
Tork W6 Basic Papers
Tork W6 Basic Papers
 :  Essity
New Product
undefined
Tork W6 Basic Papers
Supplier:  Essity
New Product
Product Information
  •  
    Colour
    L×W
    Packed
  • Wipers, refill, universal, compatible with Tork W6 dispensers, Core int.Ø: 38 mm
    Wipers, refill, universal, compatible with Tork W6 dispensers, Core int.Ø: 38 mm
    SCAA130008
    130008
    Blue
    250000×195 mm
    6 rolls (6 × 250 m)
    1 * 6 SZT
    1 * 6 SZT - 452 zł
    452 zł
    1 * 6 SZT
       452 zł
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • Wipers, refill, universal, compatible with Tork W6 dispensers, Core int.Ø: 38 mm
    Wipers, refill, universal, compatible with Tork W6 dispensers, Core int.Ø: 38 mm
    SCAA130007
    130007
    White
    250000×195 mm
    6 rolls (6 × 250 m)
    1 * 6 SZT
    1 * 6 SZT - 425 zł
    425 zł
    1 * 6 SZT
       425 zł
    New Product
    Specifications
    Environmentally Preferable
    New Product