Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Perystaltyczne pompy dozujące, perimatic premier

Dostawca: Jencons
The Perimatic Premier is a heavy-duty, microprocessor-controlled precision dispensing pump, ideally suited for rapidly and reproducibly filling a variety of containers including, vials, bottles, test tubes and petri dishes without splashing or frothing. It features manual dispensing by hand or footswitch, and automatic dispensing with interval timer. The Perimatic Premier can be used with a XY robotic module for automated filling of multiple containers.

  • Dispensing range: 0,5 to 9999,9 ml
  • Accuracy: ±1,0% for volumes over 20 ml; Repeatability:<1%
  • Microprocessor calibration
  • Memory to store calibration values, volumes and set-ups for various tubing sizes
  • Sequential slow/fast/slow dispensing

The Premier achieves its high performance due to its precision rotor and pinch valve design. The Perimatic™ Premier is particularly suited to media dispensing as its 'stand-by' mode ensures constant re-circulation of the reservoir contents, thereby keeping the media in suspension when not actively dispensing.

Informacja o dostawie Supplied with three sets of tubing assemblies and a footswitch.

Uwaga: * Dependant upon tube bore and selected speed. Based on dispensing water at 21 °C
** Tube wall thickness: 2 mm



 

ZAMÓW

Rozwiń / Zwiń
Opis Ilość Numer katalogowy VWR Ilość
Pump dispenser, Perimatic Premier 1 JENC256-005BOM
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Zakres prędkości (obr./min) 2,1 - 33,3 ml/sec*
Int.Ø tubing (mm) 8/3 mm (Max./Min.)**
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość