Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Bezprzewodowe systemy monitoringu, Smart-Vue

Dostawca: Thermo Scientific
Smart-Vue™
Ambient measurements or monitoring of laboratory equipment.

  • Easy to read display of monitored setpoints directly on module
  • Wireless data transmission (868 MHz): No wiring or rebuilding required for installation
  • Fully integrated open software: Data can easily be exported to Excel, Word or PDF
  • Safety signal transfer: Automatic signal recognition makes it possible to use each module as repeater
  • Accessible anywhere via the internet with Smart-Vue client software

These instruments safeguard the integrity of precious samples by continuously monitoring and reporting critical conditions, such as temperature, relative humidity, CO₂ percentage and differential pressure. They can be used in highly controlled environments, pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories, as well as in storage areas. If high/low setpoints are reached, notifications are immediately sent to the user by means of the chosen alert method (visual siren, fax, telephone, email or text messaging).

Certyfikaty: Traceability to 21 CFR part 11

Informacje do zamówienia More modules for monitoring CO₂, pressure, mA analogue output signals and dry contact inputs are available. Please contact your local VWR representative for further information on models and additional accessories.

Informacja o dostawie Supplied with a module, sensor, 3,6 V lithium battery, mounting holder with Velcro, magnet & hardware, CD ROM with software drivers and user manuals. Repeater (optional), receiver (required) and alarm systems need to be ordered separately.

Uwaga: * Alarm system equipped with integrated 3,6 V lithium battery for memory back-up, prevents loss of data when power fails.



 

ZAMÓW

Rozwiń / Zwiń

Akcesoria
Opis Zakres Do Waga Szer.xGłęb.xWys. Ilość Numer katalogowy VWR Ilość
Module with external PT 100 sensor (100×3 mm) calibrated at −80; −40; 0 °C −100…+150 °C Ultra-low freezers 0,43 kg 80×230×240 mm 1 620-1890
Module with external PT100 sensor (150×3 mm) calibrated at −196; −80; 0 °C −200…+50 °C LN₂ and cryogenics 0,39 kg 80×230×240 mm 1 620-1891
Module with external PT 100 sensor (150×3 mm) calibrated at 100; 200; 350 °C 100…350 °C Ovens 0,41 kg 80×230×240 mm 1 620-1892
Module with external sensor calibrated at −30; −15; 0; 20; 40 °C −40…+80 °C Single door refrigerators, freezers, cold rooms 0,37 kg 80×230×240 mm 1 620-1893
Module with external sensor calibrated at −30; −15; 0; +20; +40 °C −40…+80 °C Refrigerators, cold rooms and freezers 0,40 kg 80×230×240 mm 1 620-1875
Module with external sensor calibrated at 30; 60; 80; 100; 105 °C −40…+120 °C Incubators and water baths 0,36 kg 80×230×240 mm 1 620-1894
Module with internal sensor calibrated at 23 °C 10…50 °C Ambient temperatures 0,30 kg 80×230×240 mm 1 620-1895
Module with external dual sensor calibrated at 23 °C and 50 % RH −40…+100 °C;
0 - 100 % RH
Incubators and stability cabinets 0,35 kg 80×230×240 mm 1 620-1876
Module with differential pressure sensor calibrated at 0 to 100 Pa with 10 Pa steps −500…+500 Pa Cleanrooms 0,32 kg 60×160×260 mm 1 620-1896
Module with sensor for monitoring 4-20mA analog output signal Calibrated on 5 points: 4, 8, 12, 16 and 20mA - Any lab equipment with an available 4-20 MA analog output signal - - 1 THBRSV209-101
Module with external temperature and CO₂ sensor calibrated at 37 °C, 5 % CO₂ 0…50 °C;
0 - 10 % CO₂
CO₂ incubators - - mm 1 THBRSV210-101
Module with external PT 100 sensor (100×3 mm) not calibrated −200…+50 °C Ultra-low freezers - - 1 THBRSV200-201
Module with external PT100 sensor (150×3 mm) not calibrated 100…350 °C LN₂ and cryogenics - - 1 THBRSV201-201
Module with external PT 100 sensor (150×3 mm) not calibrated −40…+80 °C Ovens - - 1 THBRSV202-201
Module with external sensor (not calibrated) −40…+80 °C Single door refrigerators, freezers, cold rooms - - 1 THBRSV203-201
Module with external sensor (not calibrated) −40…+80 °C Refrigerators, cold rooms and freezers - - 1 THBRSV204-201
Module with internal sensor (not calibrated) 10…50 °C Ambient temperatures - - 1 THBRSV206-201
Module with external dual sensor (not calibrated) −40…+100 °C;
0 - 100 % RH
Incubators and stability cabinets - - 1 THBRSV207-201
Module with sensor for monitoring 4-20mA analog output signal not calibrated - Any lab equipment with an available 4-20 MA analog output signal - - 1 THBRSV209-201
Module with external temperature and CO₂ sensor (not calibrated) 0…50 °C;
0 - 10 % CO₂
CO₂ incubators - - mm 1 THBRSV210-201
Module with dry contact input (not calibrated) - Any lab equipment with dry alarm contacts - - 1 THBRSV212-501
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

AKCESORIA

Rozwiń / Zwiń

Accessories
Opis Do Ilość Numer katalogowy VWR Ilość
Power supply, 100/240v to 6 VDC CO2 monitering system 1 THBRSV502-500
Replacement batteries (2) - 1 THBRSV503-500-LSB
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość